Learn More
Hamilton™ Syringe Guide for Microliter™ and Gaslight™ Syringes
Designed to work with 700/7000 series MICROLITER syringes and 1700 series GASTIGHT syringes
$49.44
Specifications
Type Syringe Guide
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
14-819-20
View Documents
Hamilton™
14806
Microliter™ and Gastight™ syringes and 700 and 1700 syringe series
Each for $49.44
Add to cart
14-819-30
View Documents
Hamilton™
14906
Microliter™ and Gastight™ syringes, 7000 Series and 25-500μL capacity syringes in 700
Each for $49.44
Add to cart
Description
Description
Designed to work with 700/7000 series MICROLITER syringes and 1700 series GASTIGHT syringes
Syringe guides are designed to prevent plunger bending and the adjustable stop prevents plunger damage.
Specifications
Specifications
Syringe Guide