Learn More
Hamilton™ 900 Microliter Syringes, RN Termination, No Needle Included
The 900 series syringe is the economical version of the 800 series syringe. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 10 µL. The syringe holder is permanently attached to the syringe barrel. This reduces the initial cost of the syringe but eliminates the possibility of replacement components.
Supplier: Hamilton™ 764701
Description
- Ideal for dispensing volumes from 0.5 µL up to 10 µL
- Syringe holder is permanently attached to the syringe barrel
Specifications
5 μL |