Learn More
CD49a Hamster anti-Rat/Mouse, Brilliant Violet 711, Clone: Ha31/8, BD
Hamster Monoclonal Antibody
Manufacturer:  BD Biosciences 564863
Catalog No. BDB564863
Add to cart
Description
Description
The Ha31/8 monoclonal antibody specifically binds to the 180-kDa integrin α 1 chain (CD49a), which is a transmembrane glycoprotein that non-covalently associates with the integrin β 1 subunit (CD29) to form the α 1β 1 (complex known as VLA-1). VLA-1 has been reported to be expressed on activated T cells, monocytes, smooth muscle cells, and endothelial cells. It is a receptor for collagen and laminin. The Ha31/8 monoclonal antibody is specific for both rat and mouse CD49a. It has been reported that Ha31/8 antibody can block VLA-1-mediated binding of rat cells to collagen.
Specifications
Specifications
CD49a
Ha31/8
Brilliant Violet 711
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Hamster
IgG2, λ 1
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Itga1; Integrin alpha-1; Integrin α1; Laminin and collagen receptor; VLA-1a
Affinity Purified
Rat Emulsified Lewis Rat Glomeruli Cells
Affinity Chromatography
RUO
Primary
Murine, Rat