Learn More
Hamster anti-IgG2,λ, Isotype Control, APC-R700, Clone: Ha4/8, BD
Manufacturer:  BD Biosciences 565777
Catalog No. BDB565777
Add to cart
Description
Description
Specifications
Specifications
IgG2,λ
Ha4/8
APC-R700
Affinity Purified
Keyhole limpet hemocyanin
Affinity Chromatography
RUO
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA, protein stabilizer, glycerol, and ≤0.09% sodium azide.
Hamster
IgG2, λ
0.1mg
Store undiluted at 4°C. Do not freeze. Protect from light.
Not Provided