Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49a Hamster anti-Mouse, BUV395, Clone: Ha31/8, BD Optibuild™

Hamster Monoclonal Antibody

Manufacturer:  BD Biosciences 740262

Catalog No. BDB740262


Add to cart

Description

Description

The Ha31/8 monoclonal antibody specifically binds to the 180-kDa integrin α 1 chain (CD49a), which is a transmembrane glycoprotein that non-covalently associates with the integrin β 1 subunit (CD29) to form the α 1β 1 (complex known as VLA-1). VLA-1 has been reported to be expressed on activated T cells, monocytes, smooth muscle cells, and endothelial cells. It is a receptor for collagen and laminin. The Ha31/8 monoclonal antibody is specific for both rat and mouse CD49a. It has been reported that Ha31/8 antibody can block VLA-1-mediated binding of rat cells to collagen.

Specifications

Specifications

CD49a
Ha31/8
Brilliant Ultraviolet 395
Aqueous buffered solution containing ≤0.09% sodium azide.
Hamster
IgG2, λ1
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Itga1; Integrin alpha-1; Integrin α1; Laminin and collagen receptor; VLA-1a
Affinity Purified
Rat Emulsified Lewis Rat Glomeruli Cells
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only