Learn More
CD49a Hamster anti-Mouse, Brilliant Violet 421, Clone: Ha31/8, BD Optibuild™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 740046
Description
The Ha31/8 monoclonal antibody specifically binds to the 180-kDa integrin α 1 chain (CD49a), which is a transmembrane glycoprotein that non-covalently associates with the integrin β 1 subunit (CD29) to form the α 1β 1 (complex known as VLA-1). VLA-1 has been reported to be expressed on activated T cells, monocytes, smooth muscle cells, and endothelial cells. It is a receptor for collagen and laminin. The Ha31/8 monoclonal antibody is specific for both rat and mouse CD49a. It has been reported that Ha31/8 antibody can block VLA-1-mediated binding of rat cells to collagen.
Specifications
CD49a | |
Monoclonal | |
0.2mg/mL | |
Itga1; Integrin alpha-1; Integrin α1; Laminin and collagen receptor; VLA-1a | |
Hamster | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2 λ1 |
Flow Cytometry | |
Ha31/8 | |
Brilliant Violet 421 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat Emulsified Lewis Rat Glomeruli Cells | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only