Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hamster IgG1, λ1 Isotype Control, BV421, Clone: G235-2356, BD
Supplier: BD Biosciences 562919
Description
Host Species: Hamster
Clone: G235-2356
Isotype: IgG1, λ 1
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
G235-2356 | |
Brilliant Violet 421 | |
Trinitrophenol-KLH | |
50 μg | |
Isotype Control (Primary) | |
IgG1 λ1 |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction