Learn More
MyBioSource Lipase A, Lysosomal Acid, Hamster ELISA Kits
SDP
Supplier:  MyBioSource MBS017280
Encompass_Preferred
The MyBioSource Lipase A Lysosomal Acid Hamster ELISA Kit quantifies Lipase A in hamster biological fluids using a 96-well plate coated with a specific antibody. After binding and washing steps, a colorimetric reaction proportional to the Lipase A concentration is measured at 450 nm.
Catalog No. NC2831156
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.