Learn More
Celestron™ Hand-Held Digital Microscope Products for Science Education
Ideally suited for examining small objects such as coins, stamps and insects, as well as viewing specimen slides at low and high powers.
Manufacturer:  Celestron™ 44302C
Catalog No. S02443A
Add to cart
Includes: Metal stand that holds microscope at various positions; CD-ROM: Digital Microscope Suite; instruction manual
Description
Description
View specimens or objects on a computer screen, then take snapshot images or short videos. Examine objects such as coins, stamps, insects, gems and circuit boards.
  • Integral 2.0 MP digital camera with USB cable
  • Built-in LED illuminator
  • 10X to 40X plus 150X digital power
  • 1.3 MP digital camera for snapshots and video
  • Photo management software
  • Powered by USB 2.0 cable
  • Compatible with PC and Macintosh™
Specifications
Specifications
Education
2MP Digital
10 to 40x and 150x
6 White LED
4 oz.
2 Years
Metal stand that holds microscope at various positions, CD-ROM, digital microscope suite, instruction manual
10 to 40x and 150x
Examining small objects such as coins, stamps and insects, as well as viewing specimen slides at low and high powers
113g
USB 2 cable