Learn More
Thermo Scientific™ Hand Pump and Drain Kit for Precision™ Water Baths
Drain your Thermo Scientific™ Precision™ water bath with a hand pump or drain kit.
Manufacturer:  Thermo Scientific™ 102391
 View more versions of this product
Catalog No. 10-239-1
  Check Availability
Add to cart
Specifications
Description
Drain your Thermo Scientific™ Precision™ water bath with a hand pump or drain kit.
Hand Pump
  • Syphon to pump water out of any bath
Drain Kit
  • Quickly drain shaking, circulating, or coliform baths with this drain kit
  • Includes: Hose, quick disconnect insert, quick disconnect valve
Description & Specifications
Specifications
Hand Pump