Learn More
Purell™ Hand Sanitizing Wipes, Canister
Click to view available options
Size:
100 Canister
270 count
Description
- Durable, non-linting wipes
- Kills more than 99.99% of most common germs that may cause illness
- Removes light soils and dirt from hands
- Moisturizers keep skin soft
- Alcohol-free formulation
Specifications
Specifications
Product Type | Sanitizing Wipe |
For Use With (Application) | Removes light soils and dirt from hands. |
Size | 270 count |