Learn More
Argos Technologies™ HandE-Vac Handheld Aspirating System
A complete set of tools for aspiration—rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes with the Argos Technologies HandE-Vac Handheld Aspirator.
 View more versions of this product
Catalog No. 10-987-033
Add to cart
Description
Description
Use with Pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications Pressure-sensitive button on the top of the unit controls the specific amount of vacuum applied Operates with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems Autoclavable
Specifications
Specifications
Plastic
Multicolor
200 µL
HandE-Vac hand held aspirator
EV526
Tip Adapter
Autoclavable
Single channel plastic adapter
HandE-Vac hand held aspirator
1