Learn More
GSC Go Science Crazy Handheld Salinity Refractometer Products for Science Education
A portable kit for determining the salinity in water.
Manufacturer:  GSC Go Science Crazy REFRACT
Catalog No. S28105
Add to cart
Alert:
Not a toy. Product intended for ages 13 and up.
Description
Description
  • Easily test for salinity
  • Range of 0-28% salinity with minimum scale of 0.2%
  • Includes hard plastic case, adjustment screwdriver, dropping pipette, cleaning solution, cleaning cloth, leather carrying case, and instructions
Specifications
Specifications
Salinity refractometer
For testing water salinity
Handheld
Hard plastic case, adjustment screwdriver, dropping pipette, cleaning solution, cleaning cloth, leather carrying case, instructions