Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hourglass International™ HandPRO™ 617000 Series 16 in. Sterile Clean Class 100 Nitrile Gloves

Sterile, powder-free ambidextrous nitrile gloves compatible with ISO 5 and up.

Specifications

Color White
Length (English) 16 in.
Hand Ambidextrous
Sterility Sterile
Material Nitrile
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-423-066
View Documents
Hourglass International™
617000
X-Small Case of 160 for $465.35
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-423-067
View Documents
Hourglass International™
617001
Small Case of 160 for $465.35
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-423-068
View Documents
Hourglass International™
617002
Medium Case of 160 for $465.35
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-423-069
View Documents
Hourglass International™
617003
Large Case of 160 for $465.35
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-423-070
View Documents
Hourglass International™
617004
X-Large Case of 160 for $465.35
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-423-071
View Documents
Hourglass International™
617005
2X-Large Case of 160 for $465.35
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Sterile, powder-free ambidextrous nitrile gloves compatible with ISO 5 and up.

HandPRO™ 617000 Series 16 in. Sterile Clean Class 100 Nitrile Gloves made from premium 100% powder-free nitrile provide comfortable, elbow-length protection suitable for ISO class 5 and up controlled environments requiring a sterile glove with excellent ESD properties.
  • Sterilized by Gamma Irradiation
  • ISO 5 Compatible
  • 100% Nitrile
  • Finger-Textured
  • Ambidextrous
  • Powder-Free
  • Beaded X-Long Cuff
  • Triple Bagged

Certifications

Manufactured in an ISO 9001:2008 certified facility. Conforms to IEST-RP-CC005.4 Recommended Practice for Cleanroom Gloves

Specifications

Specifications

White
Ambidextrous
Nitrile
16 in.
Sterile
Triple Bagged
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit