Learn More
Hourglass International™ HandPRO P3™ 4511 Sterile Hand-Specific Latex Gloves
These hand-specific gloves are textured for a secure grip and feature a polymer coating for easy donning.
Manufacturer:  Hourglass International™ 451155
 View more versions of this product
Catalog No. 18-041-150
Add to cart
Description
Description
  • Sterilized by gamma irradiation
  • Hand-specific comfort
  • Easy donning
  • Low-protein latex for reduced risk of allergy
  • 12 in. extended-cuff length
  • Powder-free
  • Textured for a secure grip
  • Natural white color
  • Pair packed in a poly thermoform pouch
  • Triple-bagged in ISO 5 cleanroom environment
Specifications
Specifications
Yes
Specific
Latex
5.5
72 mm
Powder-free
Sterile
Rolled
Cleanroom, Sterile, Medical Device
Poly thermoform pouch
200 Pair/Case
0.22 mm
Natural
300 mm