Learn More
Hanging Wafer Robotic Tips: Tecan-Style Clear Tips, Presterilized
Clear tips for accurate sample delivery
Manufacturer:  Axygen Scientific TT50CHTRS
 View more versions of this product
Catalog No. 14-222-849
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Robotic Tip
Sterile
Yes
Round
Rack
Non-ANSI
Tecan™ Genesis™ Freedom™/Freedom Evo™/Miniprep with LiHa-1, 4, 8 head, Aurora Biomed Versa™ with 8-channel head
50μL
Clear
Racked
Polypropylene
96 Tips/Rack
For accurate sample delivery