Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile Available on GSA/VA Contract for Federal Government customers only.

Black tips for accurate sample delivery

Manufacturer: Axygen Scientific TT20CBKHTR

 View more versions of this product

Catalog No. 14-222-840

  Check Availability
Add to Cart

    Specifications

    Product Type Robotic Tip
    Sterility Nonsterile
    Material Polypropylene
    Quantity 96 Tips/Rack
    For Use With (Equipment) Tecan™ Genesis™ Freedom™/Freedom Evo™/Miniprep with LiHa-1, 4, 8 head, Aurora Biomed Versa™ with 8-channel head
    Autoclavable Yes
    Tip Style Racked
    Shape Round
    Packaging 96 Tips/wafer, 24 Wafers/Cs.
    Certifications/Compliance Non-ANSI
    For Use With (Application) For accurate sample delivery
    Volume (Metric) 20μL
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit