Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HAO2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113442
Description
HAO2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
HAO2 gene ontology annotations related to this gene include fatty acid oxidation.
Specifications
HAO2 | |
Polyclonal | |
Unconjugated | |
HAO2 | |
(S)-2-hydroxy-acid oxidase, peroxisomal; (S)-hydroxy-acid oxidase, peroxisomal; AI325478; Cell growth-inhibiting gene 16 protein; GIG16; glycolate oxidase; Hao2; Hao-2; Hao3; HAOX2; Haox3; hydroxyacid oxidase (glycolate oxidase) 3; hydroxyacid oxidase 2; hydroxyacid oxidase 2 (long chain); hydroxyacid oxidase 2, kidney; hydroxyacid oxidase 3 (medium-chain); hypothetical protein LOC509481; long chain alpha-hydroxy acid oxidase; Long-chain L-2-hydroxy acid oxidase; medium chain alpha-hydroxy acid oxidase; medium-chain 2-hydroxy acid oxidase HAOX3; medium-chain L-2-hydroxy acid oxidase | |
Rabbit | |
Protein G | |
RUO | |
51179, 56185, 84029 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.39 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q07523, Q9NYQ2, Q9NYQ3 | |
HAO2 | |
Recombinant Human Hydroxyacid oxidase 2 protein (2-200AA). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction