Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HAP40 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | HAP40 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300163
|
Novus Biologicals
NB300163 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB018144
|
Novus Biologicals
NB300163SS |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
HAP40 Polyclonal antibody specifically detects HAP40 in Mouse samples. It is validated for Western Blot.Specifications
HAP40 | |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
Autophagy, Neuronal Cell Markers, Neuroscience | |
PBS with 0.02% Sodium Azide | |
8263 | |
A partial peptide made to the 40 kDa mouse HAP40 protein | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Mouse | |
coagulation factor VIII-associated (intronic transcript) 1, coagulation factor VIII-associated 1, cpG island protein, DXS522E, F8A, factor VIII associated protein, factor VIII intron 22 protein, HAP40, huntingtin-associated protein 40 | |
F8A1 | |
IgG | |
Affinity Purified | |
This reacts with residues 314-325 [DGHGQDTSGQLP] of the mouse HAP40 protein (co-immunopreciptates Huntingtin protein). |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title