Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hapten 4-hydroxy-3-nitrophenyl acetyl (NP), Human anti-Non-species specific, Clone: B1-8, Novus Biologicals™

Human Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen hapten 4-hydroxy-3-nitrophenyl acetyl (NP)
Clone B1-8
Applications Western Blot, Flow Cytometry, Control
Host Species Human
Target Species Nonspecies specific
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB014300
View Documents
Novus Biologicals
NBP2810270.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB014301
View Documents
Novus Biologicals
NBP2810270.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Monoclonal antibody specifically detects hapten 4-hydroxy-3-nitrophenyl acetyl (NP) in Non-species specific Samples. It is validated for Western Blot, Flow Cytometry, Control.
Specifications

Specifications

hapten 4-hydroxy-3-nitrophenyl acetyl (NP)
Western Blot, Flow Cytometry, Control
Nonspecies specific
Primary
B1-8
Human
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit