Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Rabbit anti-Non-species specific, Alexa Fluor™ 750, Clone: B1-8, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252640AF750

Catalog No. NP22640A70


Add to cart

Description

Description

hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Monoclonal antibody specifically detects hapten 4-hydroxy-3-nitrophenyl acetyl (NP) in Non-species specific samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

hapten 4-hydroxy-3-nitrophenyl acetyl (NP)
B1-8
Western Blot, Flow Cytometry
Conjugated, Purified
hapten-protein conjugated NP12-chicken gammaglobulin (CG).
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Nonspecies specific
Flow Cytometry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Rabbit
IgG Lambda
0.1 mL
Primary
Hapten (4-hydroxy-3-nitropheny1) acetyl (NP).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit