Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Haptoglobin Antibody (HP/3837), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314068JF549
This item is not returnable.
View return policy
Description
Haptoglobin Monoclonal antibody specifically detects Haptoglobin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Haptoglobin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141, zonulin | |
| Recombinant fragment (around aa100-250) of human Haptoglobin protein (exact sequence is proprietary) (Uniprot: P00738) | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 3240 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HP/3837 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction