Learn More
Bioworld Haptoglobin from Human Plasma, 1 mg
Supplier: Bioworld 220700111
Haptoglobin from pooled human plasma Functions by binding to hemoglobin. Specifications:98.0% (HPCE), powder, color off-white Occurs as 3 major phenotypes: type 1-1, type 2-1, type 2-2.