Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haptoglobin Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI4H5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270925JF549

Catalog No. N270925J549


Add to cart

Description

Description

Haptoglobin Monoclonal antibody specifically detects Haptoglobin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Haptoglobin
OTI4H5
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
HP
Full length recombinant protein of human Haptoglobin (NP_005134) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141, zonulin
Mouse
IgG2a
0.1 mL
Primary
3240.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit