Learn More
Athens Research And Technology IncSupplier Diversity Partner HAPTOGLOBIN PHENOTYPE 1/1 LOW DFS Item
Manufacturer:  Athens Research And Technology IncSupplier Diversity Partner 16160801161/1LEL1MG
Haptoglobin, Phenotype 1-1, Human Plasma, Low Endotoxin Level: 1 MG; Lyophilized from sterile de-ionized water; MW 86,000; EC=1.20; EU<=0.1 EU/mg; Purity:> =95% by SDS-PAGE; polymorphic form. Native; Prepared and purified from human plasma.
Catalog No. 50-490-020