Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Haptoglobin Sheep anti-Human, Mouse, Rat, Porcine, Canine, Equine, Feline, Rabbit, Bovine (Negative), Chicken (Negative), Guinea Pig (Negative), Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
Supplier: Novus Biologicals NB10064425
Description
Haptoglobin Polyclonal antibody specifically detects Haptoglobin in Human, Mouse, Rat, Porcine, Canine, Equine, Feline, Rabbit, Bovine (Negative), Chicken (Negative), Guinea Pig (Negative) samples. It is validated for Immunodiffusion.Specifications
Haptoglobin | |
Polyclonal | |
Immunodiffusion Neat | |
P00738 | |
HP | |
Human haptoglobulin purified from serum | |
1 mL | |
Primary | |
Recognizes haptoglobulin, a plasma protein that has high affinity for haemoglobin. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunodiffusion | |
Unconjugated | |
Glycine Buffered Saline (pH 7.4), 0.01% Benzamidine, 0.1% EACA and 1.0mM EDTA with 0.09% Sodium Azide | |
binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141 | |
Sheep | |
IgG purified | |
RUO | |
3240 | |
Human, Mouse, Rat, Porcine, Canine, Equine, Feline, Rabbit, Bovine (Negative), Chicken (Negative), Guinea Pig (Negative) | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only