Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Haptoglobin Sheep anti-Human, Mouse, Rat, Porcine, Canine, Equine, Feline, Rabbit, Bovine (Negative), Chicken (Negative), Guinea Pig (Negative), Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
Specifications
Antigen | Haptoglobin |
---|---|
Dilution | Immunodiffusion Neat |
Applications | Immunodiffusion |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10064425
|
Novus Biologicals
NB10064425 |
1 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB017578
|
Novus Biologicals
NB100644250.25ML |
0.25 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Haptoglobin Polyclonal antibody specifically detects Haptoglobin in Human, Mouse, Rat, Porcine, Canine, Equine, Feline, Rabbit, Bovine (Negative), Chicken (Negative), Guinea Pig (Negative) samples. It is validated for Immunodiffusion.Specifications
Haptoglobin | |
Immunodiffusion | |
Unconjugated | |
Sheep | |
Human, Mouse, Rat, Porcine, Canine, Equine, Feline, Rabbit, Bovine (Negative), Chicken (Negative), Guinea Pig (Negative) | |
P00738 | |
3240 | |
Human haptoglobulin purified from serum | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunodiffusion Neat | |
Polyclonal | |
Purified | |
RUO | |
Glycine Buffered Saline (pH 7.4), 0.01% Benzamidine, 0.1% EACA and 1.0mM EDTA with 0.09% Sodium Azide | |
binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141 | |
HP | |
IgG | |
IgG purified | |
Recognizes haptoglobulin, a plasma protein that has high affinity for haemoglobin. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title