Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HARBI1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548133

Catalog No. PA548133

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and rat based on sequence homology.

Transposase-derived protein that may have nuclease activity (Potential). Does not have transposase activity.
Specifications

Specifications

HARBI1
Polyclonal
PBS with 0.09% sodium azide
Q96MB7, Q8BR93
Putative nuclease HARBI1, 31--, Harbinger transposase-derived nuclease, HARBI1, C11orf77
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
241547, 283254
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HARBI1
Liquid
HARBI1
KLH conjugated synthetic peptide between 4-33 amino acids from the N-terminal region of human HARB1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit