Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Westcott™ Hard Handle Kids Scissors

Click to view available options
Description:
Blunt
Pointed
Description
- Especially designed for a child's hand.
- Extra strong and durable stainless steel safety blades
- Molded, asymmetrical handles allow for left handed use
- Pointed scissors are most suitable for ages 4+
- Easily cut through paper, string and other craft materials
- Available in assorted colors (color can't be specified when ordered)
- Manufacturer's limited five year warranty
Specifications
Specifications
Product Type | Scissor |
Color | Assorted |
Description | Blunt |
Grade | Pre-K to 12 |
Age | 4 to 18 |
Length (English) | 5 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction