Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
John M. Maris Company Hard Rubber Spatulas
Nonsparking, wide, flexible blade
Supplier: John M. Maris Company 104
Description
Nonsparking, wide, flexible blade
- Integral handle
- Can be used with materials that affect metals adversely
- Washable
- Sterilizable
Specifications
Autoclavable | |
Spatula | |
4 in. | |
10.2 cm | |
1.9 cm | |
Integral |
Sterile | |
Designed to used with materials that affect Metals adversely, for mixing, weighing, cleaning, blending and many other uses | |
7.5 in. | |
0.75 in. | |
Hard Rubber | |
19 cm |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction