Learn More
Honeywell™ Harley-Davidson™ HD100 Series Safety Eyewear
Legendary Harley-Davidson name and logo appear on each frame and are engraved onto each lens
Manufacturer:  Honeywell™ HD102
 View more versions of this product
Catalog No. 19-053-002
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Impact-resistant polycarbonate lenses provide 99.9% UVA and UVB protection. Scratch-resistant hardcoat extends lens life. Made in the USA.
Limited Edition
  • Aerodynamic and contemporary styling
  • Wraparound design with molded-in sideshields
  • Browguards; adjustable spatulite temples
  • Silver frame
100 Series
  • Contemporary, wraparound sport styling
  • Dual-lens look; molded-in sideshields
  • Browguards; adjustable spatulite temples
  • Black frames with translucent orange accents
Harley-Davidson and its logo are registered trademarks of H-D Michigan, Inc.
Order Info
Sales of these eyewear products are restricted to businesses only; they are not available for general consumer purchase. To order, contact your Fisher Scientific Customer Service Representative.
Compliance
Meet ANSI™ Z87.1-2003 standards. The 400 series also meets CSA Z94.3-2007 standards.
Specifications
Specifications
Safety