Learn More
Honeywell™ Harley-Davidson™ HD400 Series Safety Glasses
Legendary Harley-Davidson name and logo appear on each frame and are engraved onto each lens
Manufacturer:  Honeywell™ HD402
 View more versions of this product
Catalog No. 19-053-011
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Impact-resistant polycarbonate lenses provide 99.9% UVA and UVB protection. Scratch-resistant hardcoat extends lens life. Made in the USA.
  • Wraparound nine-base lens for 180° of panoramic vision
  • Excellent side and brow coverage
  • Adjustable black, imprinted hangcord
  • Black frames
All rights reserved. Manufactured by Honeywell Safety Products under license from Harley-Davidson Motor Company.
Order Info
Sales of these eyewear products are restricted to businesses only; they are not available for general consumer purchase. To order, contact your Fisher Scientific Customer Service Representative.
Compliance
Meet ANSI Z87.1-2010 standards. The 400 series also meets CSA Z94.3 standards.
Specifications
Specifications
Polycarbonate
Impact-resistant polycarbonate
Silver Mirror
Black
Adjustable black, imprinted hangcord
Scratch-resistant, Impact-resistant