Learn More
Honeywell™ Harley-Davidson™ HD800 Series Safety Glasses
Legendary Harley-Davidson name and logo appear on each frame and are engraved onto each lens
Manufacturer:  Honeywell™ HD801
 View more versions of this product
Catalog No. 19-167-207
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Impact-resistant polycarbonate lenses provide 99.9% UVA and UVB protection. Scratch-resistant hardcoat extends lens life. Made in the USA.
  • Sleek, framelss design
  • Cross-member black and silver temples
Harley-Davidson and its logo are registered trademarks of H-D Michigan, Inc.
Order Info
Sales of these eyewear products are restricted to businesses only; they are not available for general consumer purchase. To order, contact your Fisher Scientific Customer Service Representative.
Compliance
Meet ANSI™ Z87.1-2010 standards. The 400 series also meets CSA Z94.3 standards.
Specifications
Specifications
Silver
Blue Mirror