Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harmalol Hydrochloride Hydrate 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
6028-00-8
C12H17ClN2O3
272.729
MFCD03002040
QBPGFJYXGASUTD-UHFFFAOYSA-N
hydrogen harmalol dihydrate chloride,hydrogen 1-methyl-2h,3h,4h,9h-pyrido 3,4-b indol-7-one dihydrate chloride
74764996
hydron;1-methyl-2,3,4,9-tetrahydropyrido[3,4-b]indol-7-one;chloride;dihydrate
[H+].CC1=C2C(=C3C=CC(=O)C=C3N2)CCN1.O.O.[Cl-]
6028-00-8
C12H17ClN2O3
272.729
MFCD03002040
QBPGFJYXGASUTD-UHFFFAOYSA-N
hydrogen harmalol dihydrate chloride,hydrogen 1-methyl-2h,3h,4h,9h-pyrido 3,4-b indol-7-one dihydrate chloride
74764996
hydron;1-methyl-2,3,4,9-tetrahydropyrido[3,4-b]indol-7-one;chloride;dihydrate
[H+].CC1=C2C(=C3C=CC(=O)C=C3N2)CCN1.O.O.[Cl-]
Infrared Spectrum Authentic
Quantity 1g
Water 15% max. (K.F.)
Packaging Glass bottle
Color Yellow
Assay Percent Range 98%
Formula Weight 254.72
Physical Form Crystalline Powder
Percent Purity ≥97.5% (on dry substance)
Chemical Name or Material Harmalol hydrochloride hydrate, 98%