Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harmane, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
100mg
486-84-0
C12H10N2
182.226
PSFDQSOCUJVVGF-UHFFFAOYSA-N
harman,harmane,aribine,1-methyl-9h-pyrido 3,4-b indole,locuturine,aribin,1-methyl-9h-beta-carboline,loturine,passiflorin,1-methylnorharman
5281404
CHEBI:5623
1-methyl-9H-pyrido[3,4-b]indole
CC1=NC=CC2=C1NC3=CC=CC=C23
486-84-0
C12H10N2
182.226
PSFDQSOCUJVVGF-UHFFFAOYSA-N
harman,harmane,aribine,1-methyl-9h-pyrido 3,4-b indole,locuturine,aribin,1-methyl-9h-beta-carboline,loturine,passiflorin,1-methylnorharman
5281404
CHEBI:5623
1-methyl-9H-pyrido[3,4-b]indole
CC1=NC=CC2=C1NC3=CC=CC=C23

Putative endogenous imidazoline ligand. Also MAO inhibitor

Proposed as the endogenous ligand for imidazoline binding sites. Binds to I 1 -sites in rat kidney (IC 50 = 31 nM) and I 2 -sites (K i = 49 nM). Produces dose-dependent hypotension in vivo that is reversed by efaroxan (Cat. No. 0792). Potent inhibitor of monoamine oxidase A and B (IC 50 values are 0.5 and 5 μM respectively).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 100mg
Formula Weight 182.22
Percent Purity >99%
Chemical Name or Material Harmane

Recommended Storage : Store at Room Temperature