Learn More
Harris Industries Barricade Tapes
Increase visibility of hazards to help prevent accidents
Manufacturer:  Harris Industries BT13
 View more versions of this product
Catalog No. 19-059428
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Strong, lightweight polyethylene tapes with continuous repetition of message.
  • Can be tied, stapled, or nailed into place
  • Economical and reusable
  • “ Caution” tapes have yellow background
  • “Danger” tapes have red background
Order Info
Roll size: 3 in. W x 1000 ft.L (7.6cm x 304.8m). Thickness: 4mil.
Compliance
Meet OSHA 1910.144 for marking physical hazards.
Specifications
Specifications
OSHA 1910.144
Caution Keep Out
Yellow
Caution Tape
Increase visibility of hazards to help prevent accidents
4 mil
7.62 cm
1000 ft. x 3 in. (304.8m x 7.62 cm)
Safety
HDPE
1000 ft.
304.8 m
3 in.