Harris Industries Barricade Tapes

Increase visibility of hazards to help prevent accidents

Manufacturer: HARRIS INDUSTRIES INC BT31

 View more versions of this product

Catalog No. 19-019964

  Check Availability
Add to Cart

    Specifications

    Length (English) 1000 ft.
    For Use With (Application) Increase visibility of hazards to help prevent accidents
    Length (Metric) 304.8m
    Thickness 4mil
    Width (English) 3 in.
    Width (Metric) 7.62cm
    Certification Compliance OSHA 1910.144
    Color Black, Yellow
    Displayed Text Danger Asbestos
    Material Polyethylene
    Product Type Altering Stripes Tape
    Dimensions (L x W) 1000 ft. x 3 in. (304.8m x 7.62cm)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit