Learn More
Harris Industries Barricade Tapes
Increase visibility of hazards to help prevent accidents
Manufacturer:  HARRIS INDUSTRIES INC BT12
 View more versions of this product
Catalog No. 19-019953
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1000 ft. x 3 in. (304.8m x 7.62cm)
OSHA 1910.144
Polyethylene (HDPE)
1000 ft.
304.8m
3 in.
Caution High Voltage
Yellow
Caution Tape
Increase visibility of hazards to help prevent accidents
4mil
7.62cm