Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Colored Delineator Tape

Designed for safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities.

$.94 - $41.50

Specifications

Certifications/Compliance OSHA 1910.144
Adhesive Type Rubber-based Adhesive
Material PVC
For Use With (Application) Safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities, resistance to oil, dirt and chemicals
Temperature Range (English) 14°F to 176°F
View More Specs

Products 4
Catalog Number Mfr. No. Color Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Color Dimensions (L x W) Price Quantity & Availability  
19-019996
View Documents
Harris Industries
HT200B
Black, White 18 yd. x 2 in. (16.4 m x 5.08 cm)
Each for $21.01
Add to Cart
 
19-086347
View Documents
Harris Industries
HT202BY
Black, Yellow 18 yd. x 3 in. (16.4 m x 7.62 cm)
Each for $3.69
Add to Cart
 
18-785
View Documents
Harris Industries
HT206RW
Red, White 36 yd. x 2 in. (32.9 m x 5.08 cm)
Each for $0.94
Case of 24 EA for $22.47
Add to Cart
 
19-020002
View Documents
Harris Industries
HT208BY
Black, Yellow 36 yd. x 3 in. (32.9 m x 7.62 cm)
Each for $41.50
Add to Cart
 
Description

Description

Features:

  • Alternating color stripes
  • Highly visible
  • Pressure sensitive
  • Rubber-based permanent adhesive
  • Self-wound
  • Clear laminate resists oil, dirt, chemicals and abrasion
  • 6.0mil/ 0.150mm thickness
Benefits:
  • Laminated PVC will not fade or scuff
  • Holds up to traffic and wear longer than paint
  • Can be applied quickly and easily with minimal surface preparation
  • Maintenance-free; brilliance is restored with wet mopping/washing

Red/White: Fire Protection Equipment; Black/Yellow: Physical Hazard; Black/White: Housekeeping Aisle Marking

Compliance

OSHA 1910.144

Specifications

Specifications

OSHA 1910.144
PVC
14°F to 176°F
23N/cm
Rubber-based Adhesive
Safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities, resistance to oil, dirt and chemicals
-10°C to +80°C
6 mil
Safety and Handling

Safety and Handling

ShelfLife : 1 Year

SDS
Documents

Documents

Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit