Harris Industries Colored Delineator Tape

Designed for safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities.

Manufacturer: HARRIS INDUSTRIES INC HT206RW

 View more versions of this product

Catalog No. 18-785

  Check Availability
Add to Cart

    Specifications

    Length (English) 36 yd.
    For Use With (Application) For safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities, resistance to oil, dirt and chemicals
    Shelf Life 1 Year
    Length (Metric) 32.9m
    Temperature Range (English) 14° to 176°F
    Temperature Range (Metric) - 10° to 80°C
    Tensile Strength 23N/cm
    Thickness 6mil
    Width (English) 2 in.
    Width (Metric) 5.08cm
    Adhesive Rubber based
    Certification Compliance OSHA 1910.144
    Color Red, White
    Material PVC
    Product Type Hazard Warning Tape
    Dimensions (L x W) 36 yd. x 2 in. (32.9m x 5.08cm)
    Height (Metric) 5.08cm (2 in.)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit