Learn More
Harris Industries Flagging Tape
Non-adhesive tapes available in standard or fluorescent colors
Manufacturer:  Harris Industries VST5
 View more versions of this product
Catalog No. 19-059416
Add to cart
Description
Description
  • 3cm (1.19 in.) width
  • 3.81cm (1.5 in.) core
  • Non-adhesive
  • Vinyl with monomeric flexible PVC film
  • Taffeta embossing pattern
Specifications
Specifications
Safety
PVC
300 ft.
91.4 m
2 mil
3 cm
Yellow
Flagging Tape
Marking hazards, trees, danger areas, flagging boundaries and survey markers
2620 to 2990 psi
1.19 in.
Non adhesive