Harris Industries™ Flagging Tape

Non-adhesive tapes available in standard or fluorescent colors

Manufacturer: HARRIS INDUSTRIES INC VST9

 View more versions of this product

Catalog No. 19-811-854

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Length (English) 300 ft.
    For Use With (Application) Marking hazards, trees, danger areas, flagging boundaries and survey markers
    Length (Metric) 91.4m
    Tensile Strength 2620 to 2990psi
    Adhesive Non adhesive
    Color Striped Yellow/White
    Material PVC
    Product Type Flagging Tape

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit