Harris Industries™ Flagging Tape

Non-adhesive tapes available in standard or fluorescent colors

Manufacturer: HARRIS INDUSTRIES INC VST22

 View more versions of this product

Catalog No. 19-040-024

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Marking hazards, trees, danger areas, flagging boundaries and survey markers
    Adhesive Non adhesive
    Length (English) 150 ft.
    Length (Metric) 45.72m
    Tensile Strength 2620 to 2990psi
    Thickness 4mil
    Width (English) 1.19 in.
    Width (Metric) 3cm
    Color Green
    Material PVC
    Product Type Flagging Tape

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit