Learn More
Harris Industries Hazard Warning Tape
A quick, easy and inexpensive way to mark aisles and physical hazards
Manufacturer:  Harris Industries HARHT206BY
Catalog No. 19-811-838
Add to cart
Description
Description
  • Highly visible, pressure-sensitive vinyl tape is self-wound with a permanent adhesive, and is protected with an abrasion resistant clear laminate that resists oil, dirt and chemicals
  • Colors conform to OSHA 1910.144 for marking requirements
Specifications
Specifications
OSHA 1910.144
PVC (Polyvinyl Chloride)
18 lb./in.
Physical Hazard
Hazard warning tape
Black/Yellow
Each
Designed expressly for safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities
6mil