Learn More
Harris Industries Reflective Tape
Use this engineering-grade reflective tape on poles, barricades, buildings, or other locations
Manufacturer:  Harris Industries RF2BL
 View more versions of this product
Catalog No. 19-059415
Add to cart
Description
Description
  • Pressure-sensitive adhesive back sticks to most surfaces
  • Rated for five to seven years of outdoor life, depending on conditions
  • Roll is 10 yards long (19811845 is 50 yards)
Compliance
Meets DOT specifications FP-85 and FHWA/DF-88 for Type II sheeting.
Specifications
Specifications
ASTM D4596-01
Safety
Engineering Grade Reflective
3.39 yd.
3.1 m
295 mm
5 cm
3.39 yd. x 1.96 in. (3.1m x 5 cm)
Blue
Reflective Tape
Use on poles, barricades, fences, buildings, roads, etc
69N/in.
1.96 in.
Pressure sensitive
Safety and Handling
Safety and Handling
ShelfLife : 3 Years