Learn More
Harris Industries™ Reflective Tape
Use this engineering-grade reflective tape on poles, barricades, buildings, or other locations
Manufacturer:  HARRIS INDUSTRIES INC RF2GN
 View more versions of this product
Catalog No. 19-020011
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
10 yd. x 2 in. (9.14m x 5.08cm)
Green
Reflective Tape
Use on poles, barricades, fences, buildings, roads, etc
69N/in.
2 in.
Permanent
ASTM D4596-01
Engineering Grade Reflective
10 yd.
9.14m
295mm
5.08cm
Safety and Handling
ShelfLife : 3 Years