Learn More
Harris Industries™ Reflective Tape
Use this engineering-grade reflective tape on poles, barricades, buildings, or other locations
Manufacturer:  HARRIS INDUSTRIES INC RF1RD
 View more versions of this product
Catalog No. 19-064512
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
3.39 yd. x 0.98 in. (3.1m x 2.5cm)
Red
Reflective Tape
Use on poles, barricades, fences, buildings, roads, etc
69N/in.
0.98 in.
Pressure sensitive
ASTM D4596-01
Engineering Grade Reflective
3.39 yd.
3.1m
295mm
2.5cm
Safety and Handling
ShelfLife : 3 Years