Learn More
Harris Industries™ Underground Tape
Warns excavation crews of the existence of buried pipes and cables before damage can occur
Manufacturer:  HARRIS INDUSTRIES INC UT 7
 View more versions of this product
Catalog No. 18-999-1109
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1000 ft. x 3 in. (304.8m x 7.62cm)
APWA
Polyethylene (HDPE)
1000 ft.
304.8m
4mil
7.62cm
Caution Buried Electric Line Below
Orange
Underground Tape
Warns excavation crews of the existence of buried pipes and cables before damage can occur
2100psi (transverse), 2870psi (longitudinal)
3 in.
Non adhesive