Learn More
Harris Industries™ Vinyl Plastic Delineator Tape
For color coding, aisle marking, and sealing tasks
Manufacturer:  HARRIS INDUSTRIES INC VM120WH
 View more versions of this product
Catalog No. 19-811-850
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
60 yd. x 1 in. (54.86m x 2.54cm)
White
Delineator Tape
Color coding, aisle marking, duct sealing, abrasion protection, joint sealing, pipe wrapping, decorating, light bundling, packaging, safety marking, etc
14° to 176°F
23N/cm
1 in.
Rubber based pressure sensitive
OSHA
Vinyl
60 yd.
54.86m
- 10° to 80°C
5 mil
2.54cm
Safety and Handling
ShelfLife : 1 Year