Learn More
Harris Industries™ Vinyl Plastic Delineator Tape
For color coding, aisle marking, and sealing tasks
Manufacturer:  HARRIS INDUSTRIES INC VM104 OR
 View more versions of this product
Catalog No. 18-999-1095
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
36 yd. x 3 in. (33m x 7.62cm)
Orange
Delineator Tape
Color coding, aisle marking, duct sealing, abrasion protection, joint sealing, pipe wrapping, decorating, light bundling, packaging, safety marking, etc
14° to 176°F
23N/cm
3 in.
Rubber based pressure sensitive
OSHA
Vinyl
36 yd.
33m
- 10° to 80°C
5 mil
7.62cm
Safety and Handling
ShelfLife : 1 Year